missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ceftiofur sodium, MedChemExpress
Description
Ceftiofur sodium is an antibiotic of the cephalosporin type (third generation), licensed for use in veterinary medicine.
Specifications
Specifications
| Chemical Name or Material | Ceftiofur sodium |
| Physical Form | Solid |
| CAS | 104010-37-9 |
| Color | Off-White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C19H16N5NaO7S3 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | sodium ceftiofur |
| Solubility Information | DMSO : ≥ 40 mg/mL (73.32 mM) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?