missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ceftiofur hydrochloride

Product Code. 16601842 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16601842 1 g 1g
16611842 5 g 5g
16621842 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16601842 Supplier Thermo Scientific Chemicals Supplier No. 463420010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103980-44-5
Molecular Formula C19H18ClN5O7S3
Molecular Weight (g/mol) 560.01
InChI Key KEQFDTJEEQKVLM-JUODUXDSSA-N
IUPAC Name (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-3-[(furan-2-carbonylsulfanyl)methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrochloride
SMILES Cl.CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(CSC(=O)C3=CC=CO3)=C(N2C1=O)C(O)=O)C1=CSC(N)=N1

Specifications

Melting Point >190.0°C
Quantity 1 g
Formula Weight 560.02
Chemical Name or Material Ceftiofur hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.