missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ceftiofur

Product Code. 17211182 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
17211182 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 17211182

Brand: Thermo Scientific Chemicals 465890010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Type Ceftiofur
CAS 80370-57-6
Melting Point 186.0°C to 198.0°C
Quantity 1 g
Molecular Formula C19H17N5O7S3
Molecular Weight (g/mol) 523.55
Chemical Name or Material Ceftiofur

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.