missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ceftibuten dihydrate, MedChemExpress
Description
Ceftibuten (Sch-39720) dihydrate, an antibiotic, is an orally active cephalosporin, possesses potent activity in vitro against a wide range of gram-negative and certain gram-positive pathogens.
Specifications
Specifications
| Chemical Name or Material | Ceftibuten dihydrate |
| Physical Form | Solid |
| CAS | 118081-34-8 |
| Color | Light Yellow |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C15H18N4O8S2 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | Sch-39720 dihydrate |
| Solubility Information | DMSO : 100 mg/mL (223.98 mM; Need ultrasonic) ∣H2O : 100 mg/mL (223.98 mM; ultrasonic and warming and heat to 80°C) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?