missing translation for 'onlineSavingsMsg'
Learn More

Ceftazidime, MedChemExpress

Product Code. 16402249
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16402249 1 g 1g
16461018 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16402249 Supplier Medchem Express Supplier No. HYB0593
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Ceftazidime (GR20263), an antibiotic, has a broad spectrum activity against Gram-positive and Gram-negative aerobic bacteria. Ceftazidime is also active against Enterobacteriaceae (including β-lactamase-positive strains) and is resistant to hydrolysis by most β-lactamases.

Ceftazidime (GR20263), an antibiotic, has a broad spectrum activity against Gram-positive and Gram-negative aerobic bacteria. Ceftazidime is also active against Enterobacteriaceae (including β-lactamase-positive strains) and is resistant to hydrolysis by most β-lactamases.

Specifications

Chemical Name or Material Ceftazidime
CAS 72558-82-8
Color White
Purity Grade Notes Research
Molecular Formula C22H22N6O7S2
Quantity 1 g
Synonym GR20263
Solubility Information H2O : 100 mg/mL (182.96 mM; Need ultrasonic) ∣DMSO : 25 mg/mL (45.74 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C1[C@@H](NC(/C(C2=CSC(N)=N2)=N\OC(C)(C(O)=O)C)=O)[C@@]3([H])SCC(C[N+]4=CC=CC=C4)=C(C([O-])=O)N13
Molecular Weight (g/mol) 546.58
Formula Weight 546.58
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.