missing translation for 'onlineSavingsMsg'
Learn More

Ceftazidime Hydrate (~5% Sodium Carbonate as stabilizer), TRC

Product Code. 30134670
Change view
Click to view available options
Quantity:
1 g
2.5 g
Unit Size:
1g
2.50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134670 2.5 g 2.50g
30010096 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134670 Supplier Toronto Research Chemicals Supplier No. TRCC2441282.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120618-65-7
Molecular Formula C22H22N6O7S2.x(H2O)
Molecular Weight (g/mol) 564.6
Synonym (6R,7R)-7-[(Z)-2-(Aminothiazol-4-yl)-2-(2-carboxypropoxyimino)acetamido]-3-(1-pyridiniummethyl)ceph-3-em-4-carboxylate Hydrate, Biotum Hydrate, Cefazid Hydrate, Ceftacidin Hydrate, Ceftazidine Hydrate, Fortam Hydrate, Fortaz Hydrate, Fortum Hydrate, GR 20263 Hydrate, Modacin Hydrate, Solvetan Hydrate, Tazicef Hydrate, Tazid Hydrate, Tazidime Hydrate, Tizime Hydrate, Zidim Hydrate, [6R-[6alpha,7beta(Z)]]-1-[[7-[[(2-Amino-4-thiazolyl)[(1-carboxy-1-methylethoxy)imino]acetyl]amino]-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]-pyridinium Hydroxide Inner Salt Hydrate;
IUPAC Name (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(2-carboxypropan-2-yloxyimino)acetyl]amino]-8-oxo-3-(pyridin-1-ium-1-ylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate;hydrate
SMILES O.CC(C)(O\N=C(\C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)[O-])C[n+]3ccccc3)/c4csc(N)n4)C(=O)O

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C22H22N6O7S2.H2O/c1-22(2,20(33)34)35-26-13(12-10-37-21(23)24-12)16(29)25-14-17(30)28-15(19(31)32)11(9-36-18(14)28)8-27-6-4-3-5-7-27;/h3-7,10,14,18H,8-9H2,1-2H3,(H4-,23,24,25,29,31,32,33,34);1H2/b26-13-;/t14-,18-;/m1./s1
Formula Weight 564.1097
Chemical Name or Material Ceftazidime Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.