missing translation for 'onlineSavingsMsg'
Learn More

Melford Ceftazidime Hydrate

Product Code. p-200087257
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 30018186

Brand: Melford C592005.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ceftazidime is a third-generation cephalosporin antibiotic used in research settings. It is effective against a broad spectrum of bacteria, including Gram-negative and some Gram-positive bacteria.

Ceftazidime works by inhibiting bacterial cell wall synthesis, ultimately leading to the death of susceptible bacteria. It may be utilized as a selective agent in microbiology and molecular biology experiments. Incorporate ceftazidime into culture media to selectively promote the growth of bacterial strains that carry resistance genes against this antibiotic. This is often achieved by introducing a plasmid or vector containing a ceftazidime resistance gene into the bacteria of interest.

Specifications

Chemical Name or Material Ceftazidime Hydrate
CAS 72558-82-8
Quantity 5 g
Molecular Formula C22H22N6O7S2 - xH2O
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.