missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cefradine, Mikromol™
Description
Cefradine
Specifications
Specifications
| Chemical Name or Material | Cefradine |
| CAS | 38821-53-3 |
| Analyte or Component Names | Cefradine |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C16 H19 N3 O4 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Cefradine, (6R,7R)-7-[[(2R)-amino(cyclohexa-1,4-dienyl)acetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, Cephradine, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2R)-amino-1,4-cyclohexadien-1-ylacetyl]amino]-3-methyl-8-oxo-, (6R,7R)- (9CI), Anspor, Cefradex, Cefradin, Cefrag, Cefro, Celex, Cephradin, Cesporan, Dimacef, Easkacef, Ecosporina, Eskacef, Lenzacef, Lisacef, Megacef, SQ 11436, Samedrin, Sefril, Velocef, Velosef, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2R)-2-amino-2-(1,4-cyclohexadien-1-yl)acetyl]amino]-3-methyl-8-oxo-, (6R,7R)-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[(amino-1,4-cyclohexadien-1-ylacetyl)amino]-3-methyl-8-oxo-, [6R-[6alpha,7beta(R*)]]-, 7-[D-alpha-Amino-(1,4-cyclohexadienyl)acetamide]-3-desacetoxycephalosphoranic acid, Cephalexin related, CCI 9420, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[(amino-1,4-cyclohexadien-1-ylacetyl)amino]-3-methyl-8-oxo-, [6R-[6α,7β(R*)]]-, (6R,7R)-7-[[(2R)-2-Amino-2-(1,4-cyclohexadien-1-yl)acetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[D-α-Amino-(1,4-cyclohexadienyl)acetamide]-3-desacetoxycephalosphoranic acid |
| InChI Formula | InChI=1S/C16H19N3O4S/c1-8-7-24-15-11(14(21)19(15)12(8)16(22)23)18-13(20)10(17)9-5-3-2-4-6-9/h2-3,6,10-11,15H,4-5,7,17H2,1H3,(H,18,20)(H,22,23)/t10-,11-,15-/m1/s1 |
| SMILES | CC1=C(N2[C@H](SC1)[C@H](NC(=O)[C@H](N)C3=CCC=CC3)C2=O)C(=O)O |
| IUPAC Name | (6R,7R)-7-[[(2R)-2-amino-2-cyclohexa-1,4-dien-1-ylacetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?