Learn More
Cefpirome sulfate, 95%, Thermo Scientific™
A fourth generation cephalosporin antibiotic
£72.19
Chemical Identifiers
| CAS | 98753-19-6 |
|---|---|
| Molecular Formula | C22H24N6O9S3 |
| Molecular Weight (g/mol) | 612.647 |
| MDL Number | MFCD11864966 |
| InChI Key | RKTNPKZEPLCLSF-PXPMWPIZSA-N |
| PubChem CID | 134129654 |
| IUPAC Name | (6S,7R)-7-[[2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-(6,7-dihydro-5H-cyclopenta[b]pyridin-1-ium-1-ylmethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrogen sulfate |
| SMILES | CON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC5=C4CCC5)C(=O)O.OS(=O)(=O)[O-] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 98753-19-6 | |
| 612.647 | |
| RKTNPKZEPLCLSF-PXPMWPIZSA-N | |
| (6S,7R)-7-[[2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-(6,7-dihydro-5H-cyclopenta[b]pyridin-1-ium-1-ylmethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrogen sulfate |
| C22H24N6O9S3 | |
| MFCD11864966 | |
| 134129654 | |
| CON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC5=C4CCC5)C(=O)O.OS(=O)(=O)[O-] |
Specifications
| 98753-19-6 | |
| C22H24N6O9S3 | |
| Soluble in DMSO | |
| CON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC5=C4CCC5)C(=O)O.OS(=O)(=O)[O-] | |
| 612.647 | |
| 612.66 | |
| Cefpirome sulfate |
| White | |
| MFCD11864966 | |
| RKTNPKZEPLCLSF-PXPMWPIZSA-N | |
| (6S,7R)-7-[[2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-(6,7-dihydro-5H-cyclopenta[b]pyridin-1-ium-1-ylmethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrogen sulfate | |
| 134129654 | |
| 95% |
Safety and Handling
missing translation for 'rtecsNumber' : UW8970000
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.