missing translation for 'onlineSavingsMsg'
Learn More

Cefozopran hydrochloride, MedChemExpress

Product Code. p-200097389
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16474477

Brand: Medchem Express HYB0771A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cefozopran (SCE-2787) hydrochloride is a semi-synthetic, parenteral, fourth-generation cephalosporin. Cefozopran hydrochloride, an antibiotic, has a broad spectrum of antibacterial activity, inhibiting most of the gram-negative and gram-positive organisms.

Cefozopran (SCE-2787) hydrochloride is a semi-synthetic, parenteral, fourth-generation cephalosporin. Cefozopran hydrochloride, an antibiotic, has a broad spectrum of antibacterial activity, inhibiting most of the gram-negative and gram-positive organisms.

Specifications

Chemical Name or Material Cefozopran hydrochloride
Physical Form Powder
CAS 113981-44-5
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C19H18ClN9O5S2
For Use With (Application) COVID-19-immunoregulation
Synonym SCE-2787 hydrochloride
Solubility Information DMSO : 100 mg/mL (181.16 mM; Need ultrasonic) ∣H2O : ≥ 52 mg/mL (94.20 mM)
SMILES [H]Cl.[O-]C(C(N1[C@@]2([H])[C@H](NC(/C(C3=NSC(N)=N3)=N\OC)=O)C1=O)=C(CS2)C[N+]4=C(C=CC=N5)N5C=C4)=O
Molecular Weight (g/mol) 551.99
Formula Weight 551.99
Percent Purity 95.07%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.