missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cefozopran hydrochloride, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Description
Cefozopran (SCE-2787) hydrochloride is a semi-synthetic, parenteral, fourth-generation cephalosporin. Cefozopran hydrochloride, an antibiotic, has a broad spectrum of antibacterial activity, inhibiting most of the gram-negative and gram-positive organisms.
Specifications
Specifications
| Chemical Name or Material | Cefozopran hydrochloride |
| Physical Form | Powder |
| CAS | 113981-44-5 |
| Color | Off-White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C19H18ClN9O5S2 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | SCE-2787 hydrochloride |
| Solubility Information | DMSO : 100 mg/mL (181.16 mM; Need ultrasonic) ∣H2O : ≥ 52 mg/mL (94.20 mM) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction