missing translation for 'onlineSavingsMsg'
Learn More

Cefotetan, 95+%, Thermo Scientific Chemicals

Product Code. 15509483 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15509483 1 g 1g
15519483 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15509483 Supplier Thermo Scientific Acros Supplier No. 459490010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69712-56-7
Molecular Formula C17H17N7O8S4
Molecular Weight (g/mol) 575.63
InChI Key SRZNHPXWXCNNDU-RHBCBLIFSA-N
Synonym cefotetan, cefotetanum, apacef, cefotetanum inn-latin, 6r,7s-7-4-2-amino-1-carboxy-2-oxoethylidene-1,3-dithietan-2-yl carbonyl amino-7-methoxy-3-1-methyl-1h-tetrazol-5-yl sulfanyl methyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid, cefotan tn, 6r,7s-7-4-carbamoylcarboxymethylene-1,3-dithiethane-2-carboxamido-7-methoxy-3-1-methyl-1h-tetrazol-5-yl thio methyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid, 7beta-4-2-amino-1-carboxy-2-oxoethylidene-1,3-dithietan-2-yl carbonyl amino-7alpha-methoxy-3-1-methyl-1h-tetrazol-5-yl sulfanyl methyl-3,4-didehydrocepham-4-carboxylic acid, 6r,7s-7-4-1-amino-3-hydroxy-1,3-dioxopropan-2-ylidene-1,3-dithietane-2-carbonyl amino-7-methoxy-3-1-methyltetrazol-5-yl sulfanylmethyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid
PubChem CID 53025
IUPAC Name (6R,7S)-7-[[4-(2-amino-1-carboxy-2-oxoethylidene)-1,3-dithietane-2-carbonyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)C4SC(=C(C(=O)N)C(=O)O)S4)OC)SC2)C(=O)O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: 0.51 mg/mL. Other solubilities: slightly soluble in methanol
Formula Weight 575.63
Percent Purity 95+%
Chemical Name or Material Cefotetan
compliance-icons
Product Identifier
  • Cefotetan
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.