missing translation for 'onlineSavingsMsg'
Learn More

Cefotaxime Sodium, TRC

Product Code. 30366163
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30366163 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30366163 Supplier Toronto Research Chemicals Supplier No. TRCC2429501G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cefotaxime sodium
CAS 64485-93-4
Purity Grade Notes HPLC
Molecular Formula C16 H16 N5 O7 S2 . Na
Quantity 1 g
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[(acetyloxy)methyl]-7-[[(2-amino-4-thiazolyl)(methoxyimino)acetyl]amino]-8-oxo-, monosodium salt, [6R-[6α,7β(Z)]]-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[(acetyloxy)methyl]-7-[[(2Z)-(2-amino-4-thiazolyl)(methoxyimino)acetyl]amino]-8-oxo-, monosodium salt, (6R,7R)- (9CI), (+)-Cefotaxime sodium salt, Baxim, Biolog, Biotax, Bioxim, Calcef, Califrid, CefSo, Cefacron, Cefantral, Cefcan, Cefomed, Cefomerc, Cefomerk, Cefon, Cefotam, Cefotax, Cefotaxime sodium, Cefotaxime sodium salt, Cefoxim, Cefronate, Ceftim, Ceftop, Ceftoringe, Chemcef, Clafora, Claftax, Clavox, Eaxiron, Elkotak, Epotazime, Evotaxime, Excef, Forax, Fortax, HR 756, Novatax, Oritaxim, Pretor, RU 24756, Sefagen, Sodium cefotaxime, Tolycar, Zafixime
InChI Formula InChI=1S/C16H17N5O7S2.Na/c1-6(22)28-3-7-4-29-14-10(13(24)21(14)11(7)15(25)26)19-12(23)9(20-27-2)8-5-30-16(17)18-8;/h5,10,14H,3-4H2,1-2H3,(H2,17,18)(H,19,23)(H,25,26);/q;+1/p-1/b20-9-;/t10-,14-;/m1./s1
SMILES [Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)[O-])COC(=O)C)\c3csc(N)n3
IUPAC Name sodium;(6R,7R)-3-(acetyloxymethyl)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Molecular Weight (g/mol) 477.45
Formula Weight 477.0389
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.