missing translation for 'onlineSavingsMsg'
Learn More

Cefotaxime sodium salt

Product Code. 15475009
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15475009 1 g 1g
15485009 5 g 5g
15495009 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15475009 Supplier Thermo Scientific Alfa Aesar Supplier No. J62690.03
  • ONLINE EXCLUSIVE PRICE
    £97.00
    £78.65 / 1g
    Save £18.35
    18% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cefotaxime sodium salt, CAS # 64485-93-4, is the sodium salt form of cefotaxime with β-lactamase resistant antibiotic activity. | CAS: 64485-93-4 | C16H16N5NaO7S2 | 477.44 g/mol

Cefotaxime sodium salt acts as a beta-lactamase resistant antibiotic. It is used as an effective antibacterial against gram-negative bacteria, with the notable exception of pseudomonas and penicillin-resistant strains of streptococcus pneumoniae. It is used to treat infections of the bones, joints, skin, respiratory tract and blood stream.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• The cefotaxime sodium salt is a - lactamase considered a third-generation cephalosporin with antibiotic and bactericidal activities
• This compound exerts its biological activity by binding to penicillin-binding proteins (PBP) located on the inner membrane of the bacterial cell wall leading to their inactivation. The inactivation of these proteins affects the cross-linking of peptidoglycan chains required for bacterial cell wall strength and rigidity, resulting in the weakening of cell walls and cell lysis.

Applications

• It is an efficient antibacterial compound that acts against gram-negative bacteria, without activity against gram-positive bacteria such as pseudomonas and penicillin-resistant strains of streptococcus pneumonia
• It has been used for plant cell culture and human cell lines in vitro
• Cefotaxime sodium salt has been used in antibiotic susceptibility testing (AST) methods
• Cefotaxime sodium salt is also used to remove bacteria from Ostreococcus tauri cultures

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64485-93-4
Molecular Formula C16H16N5NaO7S2
Molecular Weight (g/mol) 477.44
MDL Number MFCD00079073
InChI Key AZZMGZXNTDTSME-JUZDKLSSSA-M
PubChem CID 88631411
IUPAC Name sodium (6R,7R)-3-[(acetyloxy)methyl]-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES [Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(COC(C)=O)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1

Specifications

Type Cefotaxime Sodium Salt
Physical Form Powder
Color White to Yellow
Quantity 1 g
Beilstein 5711411
Merck Index 14,1933
Solubility Information Soluble in water.
Formula Weight 477.4
Chemical Name or Material Cefotaxime sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.