missing translation for 'onlineSavingsMsg'
Learn More

Cefotaxime, Sodium Salt, Fisher BioReagents

Product Code. 10084487
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10084487 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10084487 Supplier Fisher BioReagents Supplier No. BP29511
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Inhibitor of bacterial cell wall synthesis. Eliminates Agrobacterium species after inoculation

TRUSTED_SUSTAINABILITY

Specifications

Type Cefotaxime Sodium Salt
Physical Form Powder/Solid
CAS 64485-93-4
Loss on Drying < 3.0%
Packaging Amber Glass
Specific Rotation 58 to 64°
Concentration 916 to 964μg/mg
Quantity 1 g
Molecular Formula C16H16N5NaO7S2
MDL Number MFCD00079073
Merck Index 15, 1934
InChI Key AZZMGZXNTDTSME-JUZDKLSSSA-M
SMILES [Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(COC(C)=O)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1
IUPAC Name sodium;(6S,7R)-3-(acetyloxymethyl)-7-[[2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Molecular Weight (g/mol) 477.44
PubChem CID 88631411
Formula Weight 477.5
Percent Purity 916 to 964 μg/mg
Chemical Name or Material Cefotaxime, Sodium Salt
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cefotaxime sodium salt
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.