missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Cefotaxime
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Description
Cefotaxime Sodium Salt is a third-generation cephalosporin antibiotic.
Cefotaxime, as a cephalosporin, exerts its antibacterial effects by inhibiting bacterial cell wall synthesis. It does this by binding to penicillin-binding proteins (PBPs), disrupting the final step of peptidoglycan synthesis in the bacterial cell wall. This interference leads to structural instability and eventual lysis of the bacterial cell.
Cefotaxime sodium salt is frequently used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. Its inclusion in culture medium allows for the specific growth of bacteria lacking resistance to cefotaxime.
Specifications
Specifications
| Chemical Name or Material | Cefotaxime |
| CAS | 64485-93-4 |
| Quantity | 1 g |
| Molecular Formula | C16H16N5NaO7S2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction