missing translation for 'onlineSavingsMsg'
Learn More

Melford Cefixime Trihydrate

Product Code. 30018192
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018192 1 g 1g
30018193 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018192 Supplier Melford Supplier No. C627201.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

In the life sciences, Cefixime Trihydrate is widely utilized to study bacterial susceptibility and resistance patterns.

In the life sciences, Cefixime Trihydrate is widely utilized to study bacterial susceptibility and resistance patterns. It helps researchers investigate the effectiveness of antibiotics against various bacterial strains, contributing to a deeper understanding of microbial biology and antibiotic development.

In pharmaceutical research, Cefixime Trihydrate serves as a reference standard for antibiotic formulation and efficacy testing. Researchers rely on it to develop new antibiotics, optimize existing ones, and combat emerging antibiotic-resistant bacteria.

Specifications

Chemical Name or Material Cefixime Trihydrate
CAS 125110-14-7
Quantity 1 g
Molecular Formula C16H15N5O7S2 - 3H2O
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.