missing translation for 'onlineSavingsMsg'
Learn More

Cefixime, MedChemExpress

Product Code. 16474587
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16474587 50 mg 50mg
16451108 5 mg 5mg
16448698 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16474587 Supplier Medchem Express Supplier No. HYB1381
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cefixime is an antibiotic and a third generation cephalosporin antibiotic, useful for the treatment of a number of bacterial infections.

Cefixime is an antibiotic and a third generation cephalosporin antibiotic, useful for the treatment of a number of bacterial infections.

Specifications

Chemical Name or Material Cefixime
Physical Form Solid
CAS 79350-37-1
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C16H15N5O7S2
For Use With (Application) COVID-19-immunoregulation
Synonym FR-17027 FK-027 CL-284635
Solubility Information DMSO : 100 mg/mL (220.53 mM; Need ultrasonic)
SMILES O=C(C(N12)=C(C=C)CS[C@]2([H])[C@H](NC(/C(C3=CSC(N)=N3)=N\OCC(O)=O)=O)C1=O)O
Molecular Weight (g/mol) 453.45
Formula Weight 453.45
Percent Purity 99.44%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.