missing translation for 'onlineSavingsMsg'
Learn More

Cefiderocol, MedChemExpress

Product Code. 16497428
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497428 10 mg 10mg
16489907 5 mg 5mg
16457406 100 mg 100mg
16453457 50 mg 50mg
16443586 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16497428 Supplier Medchem Express Supplier No. HY17628

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cefiderocol (S-649266) is a siderophore cephalosporin which has a potent activity against a broad range of aerobic Gram-negative bacterial species with MIC50s of 2 μg/mL or less.

Cefiderocol (S-649266) is a siderophore cephalosporin which has a potent activity against a broad range of aerobic Gram-negative bacterial species with MIC50s of 2 μg/mL or less.

Specifications

Chemical Name or Material Cefiderocol
CAS 1225208-94-5
Color White
Purity Grade Notes Research
Molecular Formula C30H34ClN7O10S2
Quantity 10 mg
Synonym S-649266
Solubility Information DMSO : 250 mg/mL (332.35 mM; Need ultrasonic) ∣H2O : 1.06 mg/mL (1.41 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(C(N12)=C(C[N+]3(CCNC(C4=CC=C(O)C(O)=C4Cl)=O)CCCC3)CS[C@]2([H])[C@H](NC(/C(C5=CSC(N)=N5)=N\OC(C)(C(O)=O)C)=O)C1=O)[O-]
Molecular Weight (g/mol) 752.21
Formula Weight 752.21
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.