missing translation for 'onlineSavingsMsg'
Learn More

Cefepime hydrochloride monohydrate, Thermo Scientific™

Cefepime hydrochloride monohydrate, CAS # 123171-59-5, is a hydrochloride salt of the antibiotic derived from Acremonium fungus.

£84.10 - £326.00

Chemical Identifiers

CAS 123171-59-5
Molecular Formula C19H29Cl2N6O6S2+
Molecular Weight (g/mol) 572.497
MDL Number MFCD01687679
InChI Key LRAJHPGSGBRUJN-OMIVUECESA-O
PubChem CID 129893984
IUPAC Name (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-[(1-methylpyrrolidin-1-ium-1-yl)methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate;dihydrochloride
SMILES C[N+]1(CCCC1)CC2=C(N3C(C(C3=O)NC(=O)C(=NOC)C4=CSC(=N4)N)SC2)C(=O)O.O.Cl.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15487509
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
J66237.03
1g
£84.10
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15497509
SDS View Documents
Thermo Scientific Alfa Aesar
J66237.06
5g
£326.00
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description

• Cefepime hydrochloride is a fourth-generation cephalosporin antibiotic that works against most antibiotic resistant strains of bacteria

• Cefepime hydrochloride is a penicillin binding protein inhibitor that prevents peptidoglycan cross-linkage and leads to cell wall weakening and lysis

Application

• Cefepime hydrochloride monohydrate can be used for the laboratory study of microbial physiology and antibiotic resistance

• Cefepime has broad-spectrum activity; it is appropriate for research on bacteria responsible for resistant infections and physiological studies

Specifications

Chemical Identifiers

123171-59-5
572.497
LRAJHPGSGBRUJN-OMIVUECESA-O
(6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-[(1-methylpyrrolidin-1-ium-1-yl)methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate;dihydrochloride
C19H29Cl2N6O6S2+
MFCD01687679
129893984
C[N+]1(CCCC1)CC2=C(N3C(C(C3=O)NC(=O)C(=NOC)C4=CSC(=N4)N)SC2)C(=O)O.O.Cl.Cl

Specifications

123171-59-5
Light sensitive
MFCD01687679
LRAJHPGSGBRUJN-OMIVUECESA-O
(6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-[(1-methylpyrrolidin-1-ium-1-yl)methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate;dihydrochloride
129893984
Cefepime hydrochloride monohydrate
White to Yellow
C19H29Cl2N6O6S2+
Soluble in DMSO
C[N+]1(CCCC1)CC2=C(N3C(C(C3=O)NC(=O)C(=NOC)C4=CSC(=N4)N)SC2)C(=O)O.O.Cl.Cl
572.497
571.5
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c

H317-H334

missing translation for 'tsca' : No

missing translation for 'storageNote' : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.