missing translation for 'onlineSavingsMsg'
Learn More

Cefepime Dihydrochloride Monohydrate, MedChemExpress

Product Code. 16441028
Encompass_Preferred
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
This item is not returnable. View return policy

Product Code. 16441028

Brand: Medchem Express HYB0616

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cefepime Dihydrochloride Monohydrate is a broad-spectrum cephalosporin with enhanced coverage against Gram-positive and Gram-negative bacteria.

Cefepime Dihydrochloride Monohydrate is a broad-spectrum cephalosporin with enhanced coverage against Gram-positive and Gram-negative bacteria.

Specifications

Chemical Name or Material Cefepime Dihydrochloride Monohydrate
CAS 123171-59-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H28Cl2N6O6S2
Quantity 5 g
Solubility Information H2O : 10 mg/mL (17.50 mM; Need ultrasonic) ∣DMSO : 6 mg/mL (10.50 mM; Need ultrasonic)
SMILES C[N+]1(CC(CS[C@]2([H])[C@@H]3NC(/C(C4=CSC(N)=N4)=N\OC)=O)=C(C(O)=O)N2C3=O)CCCC1.[Cl-].Cl.O
Molecular Weight (g/mol) 571.5
Formula Weight 571.5
Percent Purity 99.78%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.