missing translation for 'onlineSavingsMsg'
Learn More

Cefepime Dihydrochloride Monohydrate, MedChemExpress

Product Code. 16432186
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16432186 500 mg 500mg
16442249 1 g 1g
16441028 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16432186 Supplier Medchem Express Supplier No. HYB0616500mg
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cefepime Dihydrochloride Monohydrate is a broad-spectrum cephalosporin with enhanced coverage against Gram-positive and Gram-negative bacteria.

Cefepime Dihydrochloride Monohydrate is a broad-spectrum cephalosporin with enhanced coverage against Gram-positive and Gram-negative bacteria.

Specifications

Chemical Name or Material Cefepime Dihydrochloride Monohydrate
CAS 123171-59-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H28Cl2N6O6S2
Quantity 500 mg
Solubility Information H2O : 10 mg/mL (17.50 mM; Need ultrasonic) ∣DMSO : 6 mg/mL (10.50 mM; Need ultrasonic)
SMILES C[N+]1(CC(CS[C@]2([H])[C@@H]3NC(/C(C4=CSC(N)=N4)=N\OC)=O)=C(C(O)=O)N2C3=O)CCCC1.[Cl-].Cl.O
Molecular Weight (g/mol) 571.5
Formula Weight 571.5
Percent Purity 99.78%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.