missing translation for 'onlineSavingsMsg'
Learn More

Cefazolin sodium salt

Product Code. 15476809
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15476809 5 g 5g
15466809 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15476809 Supplier Thermo Scientific Alfa Aesar Supplier No. J65274.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibits bacterial cell wall synthesis | CAS: 27164-46-1 | C14H16N8NaO4S3 | 479.504 g/mol

Inhibits bacterial cell wall synthesis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27164-46-1
Molecular Formula C14H16N8NaO4S3
Molecular Weight (g/mol) 479.504
MDL Number MFCD00056883
InChI Key MTIAAUXSENDLGW-SLNAEPSVSA-N
Synonym Cefamedin
PubChem CID 131673922
IUPAC Name (6R,7R)-3-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanylmethyl]-8-oxo-7-[[2-(tetrazol-1-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;molecular hydrogen;sodium
SMILES [HH].CC1=NN=C(S1)SCC2=C(N3C(C(C3=O)NC(=O)CN4C=NN=N4)SC2)C(=O)O.[Na]

Specifications

Type Cefazolin Sodium Salt
Physical Form Powder
Melting Point 170°C to 172°C
Color White
Quantity 5 g
Sensitivity Light sensitive
Beilstein 3585038
Merck Index 14,1917
Formula Weight 476.48
Chemical Name or Material Cefazolin sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.