missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cefazolin sodium salt, 98%

Product Code. 15344998 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 15344998

Brand: Thermo Scientific Chemicals 455210050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cefazolin sodium, CAS # 27164-46-1, is the sodium salt of the beta-lactam antibiotic cefazolin, a bactericidal compound of the cephalosporin family.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Cefazolin sodium is a beta-lactam antibiotic of the cephalosporin family

Applications

• Cefazolin binds and inactivates penicillin-binding proteins (PBP) and interferes with peptidoglycan chain crosslinking

• PBP inactivation leads to destruction of the cell wall and lysis

• Cefazolin exhibits broad spectrum activity against gram-positive and gram-negative bacteria

TRUSTED_SUSTAINABILITY

Specifications

Type Cefazolin Sodium Salt
CAS 27164-46-1
Melting Point 170°C to 177°C
Odor Odorless
Color White to Yellow
Quantity 5 g
Molecular Formula C14H16N8NaO4S3
MDL Number MFCD0005688
InChI Key MTIAAUXSENDLGW-SLNAEPSVSA-N
SMILES [HH].CC1=NN=C(S1)SCC2=C(N3C(C(C3=O)NC(=O)CN4C=NN=N4)SC2)C(=O)O.[Na]
IUPAC Name (6R,7R)-3-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanylmethyl]-8-oxo-7-[[2-(tetrazol-1-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;molecular hydrogen;sodium
Molecular Weight (g/mol) 479.504
PubChem CID 131673922
Formula Weight 476.48
Percent Purity 98%
Chemical Name or Material Cefazolin sodium salt
Show More Show Less
compliance-icons
Product Identifier
  • Cefazolin sodium salt
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.