missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cefazolin sodium salt, 98%

Product Code. 15334998 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15334998 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15334998 Supplier Thermo Scientific Chemicals Supplier No. 455210010
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Type Cefazolin Sodium Salt
CAS 27164-46-1
Melting Point 170.0°C to 177.0°C
Infrared Spectrum Authentic
Specific Rotation −17° to −20° (20°C, 589nm) (c=1,H2O)
Water 6% max.
Quantity 1 g
Molecular Formula C14H16N8NaO4S3
Solubility Information Solubility in water: 50g/L. Other solubilities: slightly soluble in methanol and ethanol, insoluble in benzene, acetone, chloroform
InChI Key MTIAAUXSENDLGW-SLNAEPSVSA-N
SMILES [HH].CC1=NN=C(S1)SCC2=C(N3C(C(C3=O)NC(=O)CN4C=NN=N4)SC2)C(=O)O.[Na]
IUPAC Name (6R,7R)-3-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanylmethyl]-8-oxo-7-[[2-(tetrazol-1-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;molecular hydrogen;sodium
Molecular Weight (g/mol) 479.504
PubChem CID 131673922
Formula Weight 476.49
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Cefazolin sodium salt
Show More Show Less
compliance-icons
Product Identifier
  • Cefazolin sodium salt
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.