missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cefazolin Sodium, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Cefazolin Sodium
Specifications
Specifications
| Chemical Name or Material | Cefazolin Sodium |
| CAS | 27164-46-1 |
| Analyte or Component Names | Cefazolin Sodium |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C14 H13 N8 O4 S3 . Na |
| Certifications/Compliance | ISO 17034 |
| Synonym | (6R,7R)-3-[[(5-Methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-8-oxo-7-[[2-(1H-tetrazol-1-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid sodium salt, (6R,7R)-3-[[(5-Methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-8-oxo-7-[(1H-tetrazol-1-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid monosodium salt, (6R-trans)-3-[[(5-Methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-8-oxo-7-[(1H-tetrazol-1-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid monosodium salt, 3-[[(5-Methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-8-oxo-7-[2-(1H-tetrazol-1-yl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid monosodium salt, 3-[[(5-Methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-7-[2-(1H-tetrazol-1-yl)-acetamido]-3-cephem-4-carboxylic acid sodium salt, 3-[[(5-Methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-8-oxo-7-[2-(1H-tetrazol-1-yl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid sodium salt, Acef, Ancef, Atirin, Biazolina, Bor-Cefazol, Cefacidal, Cefamedin, Cefazil, Cefazina, Cefazoliin, Cefazolin sodium, Cefazolin sodium salt, Cefazoline sodium, Cefazorin DC, Cephazolin sodium, Cephazolin sodium salt, Firmacef, Gramaxin, Kefzol, Lampocef, Liviclina, Monosodium cafazolin, NSC 291561, Reflin, SKF 41558, Sodium CEZ, Sodium cefazolin, Sodium cephazolin, Totacef, Zolicef, Sodium (6R,7R)-3-[[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanyl]methyl]-8-oxo-7-[(1H-tetrazol-1-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
| InChI Formula | InChI=1S/C14H14N8O4S3.Na/c1-6-17-18-14(29-6)28-4-7-3-27-12-9(11(24)22(12)10(7)13(25)26)16-8(23)2-21-5-15-19-20-21;/h5,9,12H,2-4H2,1H3,(H,16,23)(H,25,26);/q;+1/p-1/t9-,12-;/m1./s1 |
| SMILES | [Na+].Cc1nnc(SCC2=C(N3[C@H](SC2)[C@H](NC(=O)Cn4cnnn4)C3=O)C(=O)[O-])s1 |
| IUPAC Name | sodium;(6R,7R)-3-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanylmethyl]-8-oxo-7-[[2-(tetrazol-1-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction