missing translation for 'onlineSavingsMsg'
Learn More

Cefazolin sodium, MedChemExpress

Product Code. 16482246
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482246 500 mg 500mg
16428596 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16482246 Supplier Medchem Express Supplier No. HYB1078
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cefazolin sodium is a first-generation cephalosporin antibiotic and can be used in varieties of bacterial infections research. Cefazolin sodium has anti-inflammatory effect and can attenuate post-operative cognitive dysfunction (POCD).

Cefazolin sodium is a first-generation cephalosporin antibiotic and can be used in varieties of bacterial infections research. Cefazolin sodium has anti-inflammatory effect and can attenuate post-operative cognitive dysfunction (POCD).

Specifications

Chemical Name or Material Cefazolin sodium
Physical Form Solid
CAS 27164-46-1
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C14H13N8NaO4S3
For Use With (Application) COVID-19-immunoregulation
Synonym Cephazolin sodium
Solubility Information H2O : ≥ 100 mg/mL (209.87 mM) ∣DMSO : 100 mg/mL (209.87 mM; Need ultrasonic)
SMILES O=C(C(N12)=C(CSC3=NN=C(C)S3)CS[C@]2([H])[C@H](NC(CN4N=NN=C4)=O)C1=O)O[Na]
Molecular Weight (g/mol) 476.49
Formula Weight 476.49
Percent Purity 98.13%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.