missing translation for 'onlineSavingsMsg'
Learn More

Cefazolin Sodium, LoGiCal

Product Code. 30462324
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30462324

Brand: LoGiCal LGCFOR0238.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cefazolin Sodium
CAS 27164-46-1
Analyte or Component Names Cefazolin Sodium
Shipping Condition Ice pack (-20°C)
Molecular Formula C14 H13 N8 O4 S3 . Na
Certifications/Compliance ISO 9001
Synonym (6R,7R)-3-[[(5-Methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-8-oxo-7-[[2-(1H-tetrazol-1-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid sodium salt, (6R,7R)-3-[[(5-Methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-8-oxo-7-[(1H-tetrazol-1-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid monosodium salt, (6R-trans)-3-[[(5-Methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-8-oxo-7-[(1H-tetrazol-1-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid monosodium salt, 3-[[(5-Methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-8-oxo-7-[2-(1H-tetrazol-1-yl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid monosodium salt, 3-[[(5-Methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-7-[2-(1H-tetrazol-1-yl)-acetamido]-3-cephem-4-carboxylic acid sodium salt, 3-[[(5-Methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-8-oxo-7-[2-(1H-tetrazol-1-yl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid sodium salt, Acef, Ancef, Atirin, Biazolina, Bor-Cefazol, Cefacidal, Cefamedin, Cefazil, Cefazina, Cefazoliin, Cefazolin sodium, Cefazolin sodium salt, Cefazoline sodium, Cefazorin DC, Cephazolin sodium, Cephazolin sodium salt, Firmacef, Gramaxin, Kefzol, Lampocef, Liviclina, Monosodium cafazolin, NSC 291561, Reflin, SKF 41558, Sodium CEZ, Sodium cefazolin, Sodium cephazolin, Totacef, Zolicef, Sodium (6R,7R)-3-[[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanyl]methyl]-8-oxo-7-[(1H-tetrazol-1-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
InChI Formula InChI=1S/C14H14N8O4S3.Na/c1-6-17-18-14(29-6)28-4-7-3-27-12-9(11(24)22(12)10(7)13(25)26)16-8(23)2-21-5-15-19-20-21;/h5,9,12H,2-4H2,1H3,(H,16,23)(H,25,26);/q;+1/p-1/t9-,12-;/m1./s1
Solution Type Neat
SMILES [Na+].Cc1nnc(SCC2=C(N3[C@H](SC2)[C@H](NC(=O)Cn4cnnn4)C3=O)C(=O)[O-])s1
IUPAC Name sodium;(6R,7R)-3-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanylmethyl]-8-oxo-7-[[2-(tetrazol-1-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Industry Type Clinical Testing
Molecular Weight (g/mol) 476.49
Quantity 10 mg
Formula Weight 476.012 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.