missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ceefourin 1
Description
Ceefourin 1 is a selective MRP (multidrug resistance-associated protein) 4 inhibitor (IC50 = 2.6 μM). Selective for MRP4 over P-gp, MRP1 and ABCG2. Sensitizes MRP4 over-expressing cells to SN 38 (Cat. No. 2684). Ceefourin 1 inhibits cAMP efflux from acute myeloid leukemia cells (AML) and promotes apoptosis in combination with Histamine (Cat. No. 3545).
Specifications
Specifications
| Chemical Name or Material | 5-[(2-Benzothiazolylthio)methyl]-2,4-dihydro-4-methyl-3H-1,2,4-triazole-3-thione |
| CAS | 315702-40-0 |
| Quantity | 50 mg |
| Target | Multidrug Transporter Inhibitors |
| Molecular Formula | C11H10N4S3 |
| Formula Weight | 294.42 |
| Purity | >98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?