missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Calibrators

Codice prodotto. 17302743 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Description:
CEDIA Antiepileptic TDM Multi-Calibrators
Detectable Analytes:
Carbamazepine, Phenobarbital, Phenytoin, Theophylline, Valproic Acid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Description Detectable Analytes
17302743 CEDIA Antiepileptic TDM Multi-Calibrators Carbamazepine, Phenobarbital, Phenytoin, Theophylline, Valproic Acid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 17302743 Fornitore Thermo Scientific™ N. del fornitore 100007
 Restricted Item
Estimated Shipment: 29-12-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Save time while maintaining accuracy with economical Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Calibrators, optimized for use with CEDIA TDM Assays.

Streamline testing with Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Calibrators, a single calibrator kit that can be used with multiple CEDIA TDM assays. These economical kits are valued for their accuracy, precision and lot-to-lot dependability. They are available for cardiac or antiarrhythmic, antiepileptic and antibiotic drug monitoring in human serum and plasma. Each kit includes both high and low calibrators.

  • Proven technology
  • Liquid ready-to-use: no reconstitution needed
  • Multiple drug constituents per calibration kit
  • Positive linear calibration curve: the higher the absorbance reading, the more drug present
  • Excellent correlation to reference methods
  • Recombinant DNA technology provides lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Specifica

Content And Storage 2°C to 8°C
Description CEDIA Antiepileptic TDM Multi-Calibrators
Detectable Analytes Carbamazepine, Phenobarbital, Phenytoin, Theophylline, Valproic Acid
Sample Type Plasma, Serum
Type Calibrator
Quantity Low: 2 x 7.5 mL; High: 2 x 5.0 mL
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.