missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CEDIA™ Mycophenolic Acid Immunoassays, Calibrators and Controls

Complete your ISD panel with the Thermo Scientific™ CEDIA™ Mycophenolic Acid Immunoassay.

Brand:  Thermo Scientific™ 100277

Additional Details : Weight : 0.09000kg

 View more versions of this product

Product Code. 17342743

  • £275.00 / Each
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

Complete your immunosuppressant drug testing panel with the Thermo Scientific™ CEDIA™ Mycophenolic Acid Immunoassay. Like our other ISD assays, the CEDIA Mycophenolic Acid Immunoassay is accurate and precise, providing users with confidence in their results. Applications are available on our flexible and intuitive Indiko™ and Indiko Plus clinical and specialty chemical systems as well as a wide variety of other laboratory instrumentation.

Accurate:
  • Excellent low-end sensitivity
  • Correlation with gold standard methods
  • Precise throughput standard curve assay range
  • Lot-to-lot consistency
Specifications

Specifications

2°C to 8°C
Mycophenolic Acid
Calibrator
CEDIA Mycophenolic Acid Calibrator Set
Plasma
5 mL, 2 each level
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers