missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CEDIA™ Mycophenolic Acid Immunoassays, Calibrators and Controls

Product Code. 17332743 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
CEDIA Mycophenolic Acid Assay
CEDIA Mycophenolic Acid Calibrator Set
MAS Mycophenolic Acid Control 1
MAS Mycophenolic Acid Control 2
MAS Mycophenolic Acid Control 3
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description
17332743 CEDIA Mycophenolic Acid Assay
17342743 CEDIA Mycophenolic Acid Calibrator Set
17352743 MAS Mycophenolic Acid Control 1
17362743 MAS Mycophenolic Acid Control 2
17372743 MAS Mycophenolic Acid Control 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17332743 Supplier Thermo Scientific™ Supplier No. 100276
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Complete your ISD panel with the Thermo Scientific™ CEDIA™ Mycophenolic Acid Immunoassay.

Complete your immunosuppressant drug testing panel with the Thermo Scientific™ CEDIA™ Mycophenolic Acid Immunoassay. Like our other ISD assays, the CEDIA Mycophenolic Acid Immunoassay is accurate and precise, providing users with confidence in their results. Applications are available on our flexible and intuitive Indiko™ and Indiko Plus clinical and specialty chemical systems as well as a wide variety of other laboratory instrumentation.

Accurate:
  • Excellent low-end sensitivity
  • Correlation with gold standard methods
  • Precise throughput standard curve assay range
  • Lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description CEDIA Mycophenolic Acid Assay
Detectable Analytes Mycophenolic Acid
Sample Type Plasma
Type Reagent
Quantity 26 mL, 11 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.