missing translation for 'onlineSavingsMsg'
Learn More

Safety First Aid Cederroth Eye Wash with Wall Bracket and Sign

Product Code. 31759413
Change view
Click to view available options
Capacity (Metric):
500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
31759413 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31759413 Supplier Safety First Aid Supplier No. CD17
  • ONLINE EXCLUSIVE PRICE
    £25.04
    £22.40 / Pack of 1
    Save £2.64
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cederroth Wall Bracket is a robust and practical wall bracket for Cederroth Eye Wash 500 ml (REF CD15).

  • The wall bracket gives the bottle a fixed location so that is easier to identify and recognise.
  • The eye wash bottle is held in the Cederroth Wall Bracket, and opens automatically when twisted out of the bracket - so flushing can begin immediately.
  • Cederroth Wall Bracket comes with a fluorescent instruction sign.
  • Bottle of Cederroth eye wash included.

Specifications

Capacity (Metric) 500 mL
Product Type Eyewash and Shower Fixture Accessories
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.