Learn More
Wall Bracket for Cederroth Eye Wash Bottle
Product Code. 12700546
Change view
Click to view available options
For Use With (Equipment):
Cederroth Eye Wash Bottle
Product Code. For Use With (Equipment)
12700546 Cederroth Eye Wash Bottle
1 options
Product Code. 12700546 Supplier Safety First Supplier No. CD16
  • ONLINE EXCLUSIVE PRICE
    £14.06
    £8.95 / Pack of 1
    Save £5.11
    36% Off
Estimated Shipment: 24-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A robust and practical wall bracket for Cederroth Eye Wash 500 mL.
  • The wall bracket gives the bottle a fixed location so that is easier to identify and recognize
  • The eye wash bottle is held in the Cederroth Wall Bracket, and opens automatically when twisted out of the bracket, so flushing can begin immediately
  • The wall bracket is supplied empty; wash bottle is sold separately
Specifications
Capacity (Metric) 500 mL
For Use With (Equipment) Cederroth Eye Wash Bottle
Includes Comes with a Fluorescent Instruction sign
Product Type Wall Bracket