missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CECR2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £358.00
Specifications
| Antigen | CECR2 |
|---|---|
| Dilution | Western Blot 1:200-1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
CECR2 Polyclonal antibody specifically detects CECR2 in Mouse, Rat samples. It is validated for Western BlotSpecifications
| CECR2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Immunology | |
| PBS with 50% glycerol, pH7.3. | |
| 27443 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| cat eye syndrome chromosome region, candidate 2, KIAA1740 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 170-370 of human CECR2 (NP_113601.2). VQGKSNGELSLSRESEGQKNVSSIPGKTGKRRGRPPKRKKLQEEILLSEKQEENSLASEPQTRHGSQGPGQGTWWLLCQTEEEWRQVTESFRERTSLRERQLYKLLSEDFLPEICNMIAQKGKRPQRTKAELHPRWMSDHLSIKPVKQEETPVLTRIEKQKRKEEEEERQILLAVQKKEQEQMLKEERKRELEEKVKAVEG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title