missing translation for 'onlineSavingsMsg'
Learn More

Cebranopadol, MedChemExpress

Product Code. 16439177
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16439177

Brand: Medchem Express HY15536

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cebranopadol is an analgesic NOP and opioid receptor agonist with Kis/EC50s of 0.9 nM/13 nM, 0.7 nM/1.2 nM, 2.6 nM/17 nM, 18 nM/110 nM for human NOP, MOP, KOP and delta-opioid peptide (DOP) receptor, respectively.

Cebranopadol is an analgesic NOP and opioid receptor agonist with Kis/EC50s of 0.9 nM/13 nM, 0.7 nM/1.2 nM, 2.6 nM/17 nM, 18 nM/110 nM for human NOP, MOP, KOP and delta-opioid peptide (DOP) receptor, respectively.

Specifications

Chemical Name or Material Cebranopadol
CAS 863513-91-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H27FN2O
Quantity 5 mg
Synonym GRT6005
Solubility Information DMSO : 6.67 mg/mL (17.62 mM; Need ultrasonic)
SMILES CN(C)[C@]1(C2=CC=CC=C2)CC[C@]3(C(N4)=C(CCO3)C5=C4C=CC(F)=C5)CC1
Molecular Weight (g/mol) 378.48
Formula Weight 378.48
Percent Purity 96.91%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.