missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEBPZ Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25612-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene belongs to the CEBP family. The encoded protein plays a role in cellular response to environmental stimuli through a transcriptional process that involves heat shock factors, conserved DNA elements (heat shock elements or HSEs) and CCAAT boxes. The protein acts as a DNA-binding transcriptional activator and regulates the heat-shock protein 70 (HSP70) promoter in a CCAAT-dependent manner. The protein is also involved in cell growth and differentiation, particularly, hematopoietic differentiation. Methylation of the promoter of this gene or mutations within the gene may be correlated with occurrence of acute myeloid leukemia (AML).Specifications
| CEBPZ | |
| Polyclonal | |
| Unconjugated | |
| CEBPZ | |
| CBF, CBF2, CCAAT binding factor, CEBPZ, HSP CBF, NOC1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10153, 12607 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P53569, Q03701 | |
| CEBPZ | |
| CEBPZ Fusion Protein Ag11003 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction