Learn More
CEBPA Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 29388-1-AP-20UL
 View more versions of this product
Product Code. 18046675
  • £135.00 / 20µL
Description
The CEBPA antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CEBPA. This antibody recognizes human antigen. The CEBPA antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
Specifications
CEBPA
1500 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, Immunoprecipitation, ELISA
C/EBP alpha, CEBP, CEBPA
20 μL
1050
Store at -20°C. Stable for one year after shipment.