missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CEBP epsilon Antibody (PCRP-CEBPE-1G12) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27045JF549
This item is not returnable.
View return policy
Description
CEBP epsilon Monoclonal antibody specifically detects CEBP epsilon in Human samples. It is validated for Flow Cytometry,Immunocytochemistry
Specifications
| CEBP epsilon | |
| Monoclonal | |
| Janelia Fluor 549 | |
| C/EBP-epsilon, CCAAT/enhancer binding protein (C/EBP), epsilon, CCAAT/enhancer-binding protein epsilon, CRP1C/EBP epsilon | |
| Recombinant human CEBP epsilon protein | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry | |
| PCRP-CEBPE-1G12 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 1053 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction