missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEBP Delta Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41192
This item is not returnable.
View return policy
Description
CEBP Delta Polyclonal specifically detects CEBP Delta in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| CEBP Delta | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| CEBPD | |
| Antibody was raised against an 18 amino acid peptide near the carboxy terminus of human CEBPD. The immunogen is located within amino acids 170 - 220 of CEBPD. Amino Acid Squence: CKRGPDRGSPEYRQRRERN | |
| Affinity Purified | |
| RUO | |
| 1052 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 0.5-1 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 20 μg/mL | |
| C/EBP-delta, CCAAT/enhancer binding protein (C/EBP), delta, CCAAT/enhancer-binding protein delta, CELF, CRP3, NF-IL6-betaC/EBP delta, Nuclear factor NF-IL6-beta | |
| Rabbit | |
| 30 kDa | |
| 0.1 mg | |
| Primary | |
| Multiple transcript variants encoding different isoforms have been found for this gene. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction