missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEBP Delta Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92221-0.1ml
This item is not returnable.
View return policy
Description
CEBP Delta Polyclonal antibody specifically detects CEBP Delta in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| CEBP Delta | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| C/EBP-delta, CCAAT/enhancer binding protein (C/EBP), delta, CCAAT/enhancer-binding protein delta, CELF, CRP3, NF-IL6-betaC/EBP delta, Nuclear factor NF-IL6-beta | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human CEBP Delta (NP_005186.2). PAMYDDESAIDFSAYIDSMAAVPTLELCHDELFADLFNSNHKAGGAGPLELLPGGPARPLGPGPAAPRLLKREPDWGDGDAPGSLLPAQVAACAQTVVSLA | |
| 0.1 mL | |
| Cardiovascular Biology, Endocrinology, Immunology, Innate Immunity, metabolism | |
| 1052 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction