missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEBP Beta Rabbit anti-Human, Mouse, Rat, Clone: 9W4A10, Novus Biologicals™
Rabbit Monoclonal Antibody
£158.00 - £386.00
Specifications
| Antigen | CEBP Beta |
|---|---|
| Clone | 9W4A10 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18348301
|
Bio-Techne
NBP3-15761-20UL |
20 μg |
£158.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18340137
|
Novus Biologicals
NBP3-15761-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CEBP Beta Monoclonal antibody specifically detects CEBP Beta in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| CEBP Beta | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| C/EBP beta, C/EBP-beta, CCAAT/enhancer binding protein (C/EBP), beta, CCAAT/enhancer-binding protein beta, CRP2, IL6DBP, interleukin 6-dependent DNA-binding protein, LAPTCF-5, Liver activator protein, liver-enriched transcriptional activator protein, MGC32080, NF-IL6, Nuclear factor NF-IL6, nuclear factor of interleukin 6, TCF5NFIL6, Transcription factor 5 | |
| A synthetic peptide corresponding to a sequence within amino acids 246-345 of human CEBP Beta (P17676). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 9W4A10 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 1051 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title