missing translation for 'onlineSavingsMsg'
Learn More

CEBP Beta Antibody, DyLight 550, Novus Biologicals™

Product Code. 18424833 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424833 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424833 Supplier Novus Biologicals Supplier No. NBP146179R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CEBP Beta Polyclonal antibody specifically detects CEBP Beta in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEBP Beta
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias C/EBP beta, C/EBP-beta, CCAAT/enhancer binding protein (C/EBP), beta, CCAAT/enhancer-binding protein beta, CRP2, IL6DBP, interleukin 6-dependent DNA-binding protein, LAPTCF-5, Liver activator protein, liver-enriched transcriptional activator protein, MGC32080, NF-IL6, Nuclear factor NF-IL6, nuclear factor of interleukin 6, TCF5NFIL6, Transcription factor 5
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 50 and 100 of human CCAAT/Enhancer Binding Protein (C/EBP), beta using the numbering given in entry NP_005185.2 (GeneID 1051).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 1051
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.