missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEBP Beta Antibody (47A1), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04266AF488
This item is not returnable.
View return policy
Description
CEBP Beta Monoclonal specifically detects CEBP Beta in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| CEBP Beta | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CEBPB | |
| Recombinant human C/EBP-beta (1-271 aa) purified from E. coli | |
| 0.1 mL | |
| Phospho Specific | |
| 1051 | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA | |
| 47A1 | |
| Western Blot, ELISA | |
| C/EBP beta, C/EBP-beta, CCAAT/enhancer binding protein (C/EBP), beta, CCAAT/enhancer-binding protein beta, CRP2, IL6DBP, interleukin 6-dependent DNA-binding protein, LAPTCF-5, Liver activator protein, liver-enriched transcriptional activator protein, MGC32080, NF-IL6, Nuclear factor NF-IL6, nuclear factor of interleukin 6, TCF5NFIL6, Transcription factor 5 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human C/EBP beta mAb, clone 47A1, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human C/EBP beta protein. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction