missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ceapin A7

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Ceapin A7 is a selective inhibitor of ATF6α (IC50=0.59 μM). The compound selectively inhibits ATF6α over ATF6β and other branches of the unfolded protein response (UPR). Ceapin A7 inhibits ER stress-induced upregulation of BiP in vitro. Ceapin A7 inhibits cleavage and functional activation of endogenous ATF6 in response to ER stress in vitro.
Specifications
Specifications
| Chemical Name or Material | N-[1-[[2,4-bis(Trifluoromethyl)phenyl]methyl]-1 H-pyrazol-4-yl]-5-(2-furanyl)-3-isoxazolecarboxamide |
| CAS | 2323027-38-7 |
| Quantity | 10 mg |
| Target | Other ER Stress/UPR Activators |
| Molecular Formula | C20H12F6N4O3 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction