missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM8/CD66b Antibody (SR2195), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | CEACAM8/CD66b |
|---|---|
| Clone | SR2195 |
| Dilution | Western Blot 1:500-1:2000 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18619684
|
Novus Biologicals
NBP3-21965-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18638234
|
Novus Biologicals
NBP3-21965-50ul |
50 μL |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CEACAM8/CD66b Monoclonal antibody specifically detects CEACAM8/CD66b in Human samples. It is validated for Western BlotSpecifications
| CEACAM8/CD66b | |
| Western Blot 1:500-1:2000 | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| Carcinoembryonic antigen CGM6, carcinoembryonic antigen gene family member 6, carcinoembryonic antigen-related cell adhesion molecule 8, CD66b, CD66b antigen, CD67, CD67 antigen, CGM6, NCA-95, Non-specific cross-reacting antigen NCA-95 | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2195 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 1088 | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title