Learn More
CEACAM8/CD66b Antibody (24H12), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26660-100ul
 View more versions of this product
Product Code. 30043030
  • £406.00 / 100µL
Description
CEACAM8/CD66b Monoclonal antibody specifically detects CEACAM8/CD66b in Human samples. It is validated for Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CEACAM8/CD66b
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Flow Cytometry, ELISA
24H12
Flow Cytometry 1:50-1:200, ELISA
Carcinoembryonic antigen CGM6, carcinoembryonic antigen gene family member 6, carcinoembryonic antigen-related cell adhesion molecule 8, CD66b, CD66b antigen, CD67, CD67 antigen, CGM6, NCA-95, Non-specific cross-reacting antigen NCA-95
A synthesized peptide derived from Human CEACAM8/CD66b [UniProt P31997]
100 μL
Cancer
1088
Store at -20 to -70C. Avoid freeze-thaw cycles.