missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM7 Antibody (962703), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB44781P
This item is not returnable.
View return policy
Description
CEACAM7 Monoclonal antibody specifically detects CEACAM7 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, CyTOF
Specifications
| CEACAM7 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry, CyTOF | |
| 962703 | |
| Flow Cytometry, ELISA, Immunohistochemistry, CyTOF-ready | |
| Carcinoembryonic antigen CGM 2, Carcinoembryonic antigen CGM2, Carcinoembryonic antigen gene family member 2, Carcinoembryonic antigen related cell adhesion molecule 7, CEA, CEACAM 7, CGM 2, CGM2 | |
| Chinese Hamster Ovary cell line, CHO-derived recombinant human CEACAM-7, Thr36-Asn233, Accession # Q14002 | |
| 0.1 mL | |
| Cancer | |
| 1087 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction