missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CEACAM6/CD66c Antibody (tinurilimab) - Humanized, IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28575-1mg
This item is not returnable.
View return policy
Description
CEACAM6/CD66c Monoclonal antibody specifically detects CEACAM6/CD66c in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CEACAM6/CD66c | |
| Monoclonal | |
| Unconjugated | |
| P40199 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| tinurilimab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| carcinoembryonic antigen-related cell adhesion molecule 6, carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific crossreacting antigen), CD66c, CD66c antigen, NCACEAL, Non-specific crossreacting antigen, Normal cross-reacting antigen | |
| CEACAM6/ CD66c | |
| 1 mg | |
| Cancer | |
| 4680 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction