missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CEACAM6/CD66c Antibody Pair [HRP]

Product Code. 18274366 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18274366

Brand: Novus Biologicals™ NBP27928815PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CEACAM6/CD66c.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 9.34 pg/mL
Assay Sensitivity 9.34-600 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CEACAM6/CD66c.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias carcinoembryonic antigen-related cell adhesion molecule 6, carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific crossreacting antigen), CD66c, CD66c antigen, NCACEAL, Non-specific crossreacting antigen, Normal cross-reacting antigen
Gene ID (Entrez) 4680
Gene Symbol CEACAM6
Host Species Rabbit
Kit Contents
  • Standard: Each vial contains 40 ng of recombinant Human CEACAM6/CD66c.
  • Capture Antibody: 0.4 mg/mL of rabbit anti-Human CEACAM6/CD66c monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.15 mg/mL of rabbit anti-Human CEACAM6/CD66c monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Cancer
    Storage Requirements Storage is content dependent.
    Target CEACAM6/CD66c
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 600 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.